Learn More
Medchemexpress LLC RN-1665 | 1803003-65-7 | 99.93% | 513.54 | 100 MG
SDP
Catalog No. 5000395187
Encompass_Preferred
RN-1665 is an orally active TRPV4 antagonist that exhibits excellent selectivity for related TRP receptors such as TRPV1, TRPV3, and TRPM8. It serves as a TRPV4 probe for focused screens, showing IC50s of 0.26 μM for human TRPV4 and 0.39 μM for rat TRPV4.
  • Orally active.
  • TRPV4 antagonist.
  • Exhibits excellent selectivity for related TRP receptors such as TRPV1, TRPV3, and TRPM8.
  • Functions as a TRPV4 probe for focus screens.
  • Has an IC50 of 0.26 μM for human TRPV4.
  • Has an IC50 of 0.39 μM for rat TRPV4.
Catalog No. 50-003-95187 Supplier Medchemexpress LLC Supplier No. HY160900100MG
RN-1665 is an orally active TRPV4 antagonist that exhibits excellent selectivity for related TRP receptors such as TRPV1, TRPV3, and TRPM8. It serves as a TRPV4 probe for focused screens, showing IC50s of 0.26 μM for human TRPV4 and 0.39 μM for rat TRPV4.
  • Orally active.
  • TRPV4 antagonist.
  • Exhibits excellent selectivity for related TRP receptors such as TRPV1, TRPV3, and TRPM8.
  • Functions as a TRPV4 probe for focus screens.
  • Has an IC50 of 0.26 μM for human TRPV4.
  • Has an IC50 of 0.39 μM for rat TRPV4.