Learn More
TARGETMOL CHEMICALS INC RN-1734 25MG
SDP
Catalog No. 502579181
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RN-1734 is selective TRPV4 channel antagonist(IC50 of 2.3 uM5.9 uM3.2 uM for hTRPV4 mTRPV4 and rTRPV4respectively)purity: 98%
Catalog No. 50-257-9181 Supplier TARGETMOL CHEMICALS INC Supplier No. T757225MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RN-1734 is selective TRPV4 channel antagonist(IC50 of 2.3 uM5.9 uM3.2 uM for hTRPV4 mTRPV4 and rTRPV4respectively)purity: 98%