Learn More
Medchemexpress LLC HY-102014 5mg Medchemexpress, RN-18 CAS:431980-38-0 Purity:>98%
SDP
Catalog No. 501967950
Encompass_Preferred
Medchemexpress, HY-102014 5mg RN-18 CAS:431980-38-0 RN-18 is a HIV-1 viral infectivity factor (HIV-1 Vif) inhibitor with an IC50 of 6 μM in nonpermissive H9 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7950 Supplier Medchemexpress LLC Supplier No. HY1020145MG
Medchemexpress, HY-102014 5mg RN-18 CAS:431980-38-0 RN-18 is a HIV-1 viral infectivity factor (HIV-1 Vif) inhibitor with an IC50 of 6 μM in nonpermissive H9 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.