Learn More
Cayman Chemical RN-1hydrochlorIde 10mg
SDP
Catalog No. 502965101
Encompass_Preferred
A potent, irreversible inhibitor of LSD1 (IC50 = 70 nM); penetrates the blood-brain barrier and blocks long-term but not short-term memory in mice
Catalog No. 50-296-5101 Supplier Cayman Chemical Supplier No. 1812410 MG
A potent, irreversible inhibitor of LSD1 (IC50 = 70 nM); penetrates the blood-brain barrier and blocks long-term but not short-term memory in mice