Learn More
Mascon Medical Pipet Tips, RN, 200 L, Filter, Clear, Rack, Sterile, 96 pcs/rack, 50 racks/case
SDP
Catalog No. NC3527227
Encompass_Preferred
Compatibility:Rainin LTS SeriesFeaturesImported polypropylene(PP),USP Class VI standardsSmooth inner wall,reduce liquid residue,ensure the accuracy of liquid suctionDNA-free,DNase/RNase-free,Pyrogen/Endotoxin-freeUltra-grinding &molding technology,release agentfreeWithstand temperature ranging fom-80 to 121,stable perfomance underhigh temperature andhigh pressureExtrathin-wall design increase productflexibilityand improve heat transfer efficiency
Catalog No. NC3527227 Supplier Mascon Medical Supplier No. MM8600208C
Compatibility:Rainin LTS SeriesFeaturesImported polypropylene(PP),USP Class VI standardsSmooth inner wall,reduce liquid residue,ensure the accuracy of liquid suctionDNA-free,DNase/RNase-free,Pyrogen/Endotoxin-freeUltra-grinding &molding technology,release agentfreeWithstand temperature ranging fom-80 to 121,stable perfomance underhigh temperature andhigh pressureExtrathin-wall design increase productflexibilityand improve heat transfer efficiency