Learn More
Medchemexpress LLC HY-18018 5mg Medchemexpress, RN486 CAS:1242156-23-5 Purity:>98%
SDP
Catalog No. 502082012
Encompass_Preferred
Medchemexpress, HY-18018 5mg RN486 CAS:1242156-23-5 RN486 is a potent, selective and orally active Btk inhibitor with an IC50 of 4.0 nM and a Kd of 0.31 nM. RN486 is less active for other kinases. RN486 can be used for rheumatoid arthritis and systemic lupus erythematosus research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2012 Supplier Medchemexpress LLC Supplier No. HY180185MG
Medchemexpress, HY-18018 5mg RN486 CAS:1242156-23-5 RN486 is a potent, selective and orally active Btk inhibitor with an IC50 of 4.0 nM and a Kd of 0.31 nM. RN486 is less active for other kinases. RN486 can be used for rheumatoid arthritis and systemic lupus erythematosus research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.