Learn More
LGC GENOMICS LLC (Lucigen) RNA 5' Polyphosphatase, 200U @ 20 U/LDephosphorylate tri- and di-phosphorylated RNA molecules while avoiding 5'-capped and mono-phosphorylated RNAs.
Catalog No. NC0888901
Encompass
RNA 5 Polyphosphatase is a Mg2+-independent phosphohydrolase discovered and characterized by Epicentre scientists. The enzyme sequentially removes the and phosphates from 5-triphosphorylated RNA (such as primary RNA transcripts): 5 pppNOH 3 5 pNOH 3 + 2 Pi
This item is not returnable. View return policy
Catalog No. NC0888901 Supplier LGC GENOMICS LLC Supplier No. RP8092H
RNA 5 Polyphosphatase is a Mg2+-independent phosphohydrolase discovered and characterized by Epicentre scientists. The enzyme sequentially removes the and phosphates from 5-triphosphorylated RNA (such as primary RNA transcripts): 5 pppNOH 3 5 pNOH 3 + 2 Pi