Learn More
New England Biolabs, Inc. RNA 5’ Pyrophosphohydrolase (RppH) – 200 units
SDP
Catalog No. 504381
Encompass
The bacterial RNA 5' Pyrophosphohydrolase (RppH) removes pyrophosphate from the 5' end of triphosphorylated RNA to leave a 5' monophosphate RNA. The RppH protein was also known as NudH/YgdP which can split Ap5A to ADP and ATP.
Catalog No. 50-438-1 Supplier New England Biolabs, Inc. Supplier No. M0356S
The bacterial RNA 5' Pyrophosphohydrolase (RppH) removes pyrophosphate from the 5' end of triphosphorylated RNA to leave a 5' monophosphate RNA. The RppH protein was also known as NudH/YgdP which can split Ap5A to ADP and ATP.