Learn More
Medchemexpress LLC RNA, (Am-G-Cm-U-Gm-C-Am-U-Cm-A-Gm-G-Um-U-Gm-G-Cm-A-Cm), complex with RNA (G-Um-G... | 1 MG
SDP
Catalog No. 5000373188
Encompass_Preferred
PF-04523655 sodium is a siRNA directed against the RTP801 gene and acts as an inhibitor of the mammalian target of rampamycin complex 1 (mTORC1).
  • SiRNA directed against the RTP801 gene
  • Acts as an inhibitor of the mammalian target of rampamycin complex 1 (mTORC1)
  • White to off-white solid appearance
  • Purity (HPLC) of 98.71%
  • Consistent with structure (MS)
  • Product tested and complies with specifications
  • For research use only
Catalog No. 50-003-73188 Supplier Medchemexpress LLC Supplier No. HY153493A1MG
PF-04523655 sodium is a siRNA directed against the RTP801 gene and acts as an inhibitor of the mammalian target of rampamycin complex 1 (mTORC1).
  • SiRNA directed against the RTP801 gene
  • Acts as an inhibitor of the mammalian target of rampamycin complex 1 (mTORC1)
  • White to off-white solid appearance
  • Purity (HPLC) of 98.71%
  • Consistent with structure (MS)
  • Product tested and complies with specifications
  • For research use only