Learn More
Biochain Institute Inc FastHyb-Hybridization Solution, 250 ml/PK
SDP
Catalog No. 50183126
Encompass_Preferred
BioChains' FastHyb-Hybridization solution not only increases the sensitivity but also reduces the hybridization time. It is compatible with both radioactive and non-radioactive probes. This solution always results low back ground even with over night incubations.
Catalog No. 50-183-126 Supplier Biochain Institute Inc Supplier No. L1031250
BioChains' FastHyb-Hybridization solution not only increases the sensitivity but also reduces the hybridization time. It is compatible with both radioactive and non-radioactive probes. This solution always results low back ground even with over night incubations.