Learn More
Biochain Institute Inc IsHyb In Situ Hybridization (ISH) Kit for 50 slides, 1 kit/PK
SDP
Catalog No. 50183128
Encompass_Preferred
BioChain's IsHyb In Situ Hybridization Kit provides the reagents, including mRNA hybridization buffer, NBT/BCIP color development solution and washing buffer solutions, for prehybridization, hybridization, and post hybridization treatments.
Catalog No. 50-183-128 Supplier Biochain Institute Inc Supplier No. K2191050
BioChain's IsHyb In Situ Hybridization Kit provides the reagents, including mRNA hybridization buffer, NBT/BCIP color development solution and washing buffer solutions, for prehybridization, hybridization, and post hybridization treatments.