Learn More
Biochain Institute Inc Hybridization Solution, 5 ml/PK
SDP
Catalog No. 50183130
Encompass_Preferred
BioChain's IsHyb In Situ Hybridization Kit provides the reagents, including mRNA hybridization buffer, NBT/BCIP color development solution and washing buffer solutions, for prehybridization, hybridization, and post hybridization treatments.
Catalog No. 50-183-130 Supplier Biochain Institute Inc Supplier No. K21910502
BioChain's IsHyb In Situ Hybridization Kit provides the reagents, including mRNA hybridization buffer, NBT/BCIP color development solution and washing buffer solutions, for prehybridization, hybridization, and post hybridization treatments.