Learn More
Medchemexpress LLC RNA aptamer Corn (sodium) | 91.3% | 10 MG
SDP
Catalog No. 5000287168
Encompass_Preferred
RNA Aptamer Corn (sodium) is a fluorogenic 28-nucleotide RNA aptamer that binds the DFHO chromophore to produce bright red fluorescence for RNA imaging in live cells. The aptamer shows increased photostability and low cytotoxicity upon illumination. A 36-nt construct (28-nt core plus a 4-bp stem) is described for enhanced stability. Supplied as a sodium salt with reported purity of 91.29% and available in multiple pack sizes including 10 mg.
  • Enables bright red fluorescence upon binding DFHO.
  • Suitable for live-cell RNA imaging with low cytotoxicity.
  • Improved photostability versus other aptamer-chromophore complexes.
  • Provided as sodium salt with reported purity of 91.29%.
  • Includes sequence information for minimal (28 nt) and extended (36 nt) constructs.
  • Store sealed at -20°C; in solvent, store at -80°C for long-term.
Catalog No. 50-002-87168 Supplier Medchemexpress LLC Supplier No. HY15384310MG
RNA Aptamer Corn (sodium) is a fluorogenic 28-nucleotide RNA aptamer that binds the DFHO chromophore to produce bright red fluorescence for RNA imaging in live cells. The aptamer shows increased photostability and low cytotoxicity upon illumination. A 36-nt construct (28-nt core plus a 4-bp stem) is described for enhanced stability. Supplied as a sodium salt with reported purity of 91.29% and available in multiple pack sizes including 10 mg.
  • Enables bright red fluorescence upon binding DFHO.
  • Suitable for live-cell RNA imaging with low cytotoxicity.
  • Improved photostability versus other aptamer-chromophore complexes.
  • Provided as sodium salt with reported purity of 91.29%.
  • Includes sequence information for minimal (28 nt) and extended (36 nt) constructs.
  • Store sealed at -20°C; in solvent, store at -80°C for long-term.