Learn More
Zymo Research Corporation RNA Binding Buffer, 50 ml
SDP
Catalog No. 501251668
Encompass_Preferred
RNA Binding Buffer is used for efficient clean-up and concentration of RNA from any reaction (e.g., DNase I treated, in vitro transcription, etc.).
Catalog No. 50-125-1668 Supplier Zymo Research Corporation Supplier No. R1013250
RNA Binding Buffer is used for efficient clean-up and concentration of RNA from any reaction (e.g., DNase I treated, in vitro transcription, etc.).