Learn More
Enzo Life Sciences AMPINEXT™ RNA Bisulfite-Seq Kit (Illumina) (24Reactions)
SDP
Catalog No. 502013799
Encompass_Preferred
The AMPINEXT™ RNA Bisulfite-Seq Kit (Illumina) is a complete set of optimized reagents designed to carry out RNA bisulfite conversion, followed by a "post-bisulfite" library preparation process for Illumina platform-based bisulfite sequencing, all in one kit. Intended applications include whole transcriptome RNA bisulfite sequencing and various other RNA bisulfite-based next generation sequencing techniques for RNA methylation analysis. The optimized protocol and components of the kit allow the RNA to be bisulfite converted and fragmented simultaneously followed by quick non-barcoded (singleplexed) and barcoded (multiplexed) library construction using low-nanogram quantities of bisulfite converted RNA. Assay Time: 6 hours.
Catalog No. 50-201-3799 Supplier Enzo Life Sciences Supplier No. ENZGEN5100024
The AMPINEXT™ RNA Bisulfite-Seq Kit (Illumina) is a complete set of optimized reagents designed to carry out RNA bisulfite conversion, followed by a "post-bisulfite" library preparation process for Illumina platform-based bisulfite sequencing, all in one kit. Intended applications include whole transcriptome RNA bisulfite sequencing and various other RNA bisulfite-based next generation sequencing techniques for RNA methylation analysis. The optimized protocol and components of the kit allow the RNA to be bisulfite converted and fragmented simultaneously followed by quick non-barcoded (singleplexed) and barcoded (multiplexed) library construction using low-nanogram quantities of bisulfite converted RNA. Assay Time: 6 hours.