Learn More
Apexbio Technology LLC RNA Clean Beads(Synonyms: RNA purification beads, RNA magnetic beads, Magnetic RNA purification beads, RNA cleanup beads), 450ml.
SDP
Catalog No. 5000061338
Encompass_Preferred
RNA Clean Beads are magnetic microparticles designed for rapid high-throughput RNA purification from DNase-treated samples The beads function through reversible nucleic acid binding RNA molecules from 100 bp to 50 kb bind selectively to the bead surfaces upon addition of binding solution allowing separation in a magnetic field Contaminants including DNA digestion fragments primer dimers salts and residual proteins are eliminated in subsequent washing steps RNA is then eluted using sterile water or elution buffer yielding purified RNA suitable for downstream enzymatic digestion sequencing hybridization and PCR analysis This product can integrate seamlessly into automated nucleic acid purification workflows
Catalog No. 50-000-61338 Supplier Apexbio Technology LLC Supplier No. K1812450
Add to Cart
Add to Cart
Request a Quote
RNA Clean Beads are magnetic microparticles designed for rapid high-throughput RNA purification from DNase-treated samples The beads function through reversible nucleic acid binding RNA molecules from 100 bp to 50 kb bind selectively to the bead surfaces upon addition of binding solution allowing separation in a magnetic field Contaminants including DNA digestion fragments primer dimers salts and residual proteins are eliminated in subsequent washing steps RNA is then eluted using sterile water or elution buffer yielding purified RNA suitable for downstream enzymatic digestion sequencing hybridization and PCR analysis This product can integrate seamlessly into automated nucleic acid purification workflows