Learn More
Zymo Research Corporation RNA Clean & Concentrator™-100 (25 Preps) w/ Zymo-Spin™ V-E Columns w/ Reservoir (Capped)
SDP
Catalog No. 50444570
Encompass_Preferred
This 15 minute procedure is based on the use of a unique single-buffer system and Zymo-Spin™ column technology that allows for selective recovery of total RNA (> 17 nt), large RNAs (> 200 nt), and/or small RNAs (17-200 nt). The procedure is easy: Add binding buffer to your sample, then bind, wash and elute ultra-pure RNA. The RNA can be eluted from the Zymo-Spin™ V-E Column in 100 l of RNase-free water. The highly concentrated, purified RNA is suitable for all subsequent analyses and molecular manipulations.
Catalog No. 50-444-570 Supplier Zymo Research Corporation Supplier No. R1019
This 15 minute procedure is based on the use of a unique single-buffer system and Zymo-Spin™ column technology that allows for selective recovery of total RNA (> 17 nt), large RNAs (> 200 nt), and/or small RNAs (17-200 nt). The procedure is easy: Add binding buffer to your sample, then bind, wash and elute ultra-pure RNA. The RNA can be eluted from the Zymo-Spin™ V-E Column in 100 l of RNase-free water. The highly concentrated, purified RNA is suitable for all subsequent analyses and molecular manipulations.