Learn More
Zymo Research Corporation RNA Clean & Concentrator™-5 (200 Preps) w/ Zymo-Spin™ IC Columns (Capped)
SDP
Catalog No. NC0622892
Encompass_Preferred
This 5 minute procedure is based on the use of a unique single-buffer system and Zymo-Spin™ technology that allows for selective recovery of total RNA (> 17nt), large RNAs (> 200 nt), and/or small RNAs (17-200 nt).The procedure is easy: Add binding buffer and ethanol to your sample,then bind, wash and elute ultra-pure RNA. The RNA can be eluted from the Zymo-Spin™ IC Column in as little as 6 l of RNase-free water. The highly concentrated, purified RNA is suitable for all subsequent analyses and molecular manipulations.
Catalog No. NC0622892 Supplier Zymo Research Corporation Supplier No. R1016
This 5 minute procedure is based on the use of a unique single-buffer system and Zymo-Spin™ technology that allows for selective recovery of total RNA (> 17nt), large RNAs (> 200 nt), and/or small RNAs (17-200 nt).The procedure is easy: Add binding buffer and ethanol to your sample,then bind, wash and elute ultra-pure RNA. The RNA can be eluted from the Zymo-Spin™ IC Column in as little as 6 l of RNase-free water. The highly concentrated, purified RNA is suitable for all subsequent analyses and molecular manipulations.