Learn More
Zymo Research Corporation RNA Clean & Concentrator MagBead (supplied with DNaseI set) (96 preps)
SDP
Catalog No. 502037254
Encompass_Preferred
The procedure is simple: Add binding buffer and MagBinding Beads to your sample, then wash and elute ultra-pure RNA. The unique singlebuffer system and magnetic bead technology can be used on an automated platform (or manually) for high-throughput processing.
Catalog No. 50-203-7254 Supplier Zymo Research Corporation Supplier No. R1082
The procedure is simple: Add binding buffer and MagBinding Beads to your sample, then wash and elute ultra-pure RNA. The unique singlebuffer system and magnetic bead technology can be used on an automated platform (or manually) for high-throughput processing.