Learn More
Norgen Biotek Corp RNA Clean-Up and Concentration Kit, 100 Prep
Catalog No. NC1292290
Encompass_Preferred
This kit is designed to clean-up and concentrate RNA (including miRNA) from TRIzol and TRI Reagent, enzymatic reactions, in vitro transcription, labelling reactions, etc. This is a robust kit for all clean-up and concentration purposes for up to 35 µg of RNA in solution. The purified RNA is of the highest purity and integrity, and can be used in a number of downstream applications.The kit purifies all sizes of RNA, from large mRNA and ribosomal RNA down to miRNA, siRNA and lncRNA.The kit elutes RNA in 20 µL for all types of downstream applications. For smaller elution volumes of 8 µL, please refer to the microspin column kits such as the RNA Clean-Up and Concentration Micro-Elute Kit for the highest concentration of RNA sample.
Catalog No. NC1292290 Supplier Norgen Biotek Corp Supplier No. 43200
This kit is designed to clean-up and concentrate RNA (including miRNA) from TRIzol and TRI Reagent, enzymatic reactions, in vitro transcription, labelling reactions, etc. This is a robust kit for all clean-up and concentration purposes for up to 35 µg of RNA in solution. The purified RNA is of the highest purity and integrity, and can be used in a number of downstream applications.The kit purifies all sizes of RNA, from large mRNA and ribosomal RNA down to miRNA, siRNA and lncRNA.The kit elutes RNA in 20 µL for all types of downstream applications. For smaller elution volumes of 8 µL, please refer to the microspin column kits such as the RNA Clean-Up and Concentration Micro-Elute Kit for the highest concentration of RNA sample.