Learn More
INTACT GENOMICS INC RNA Cleanup Kit 100 Preps
Catalog No. 5000083005
Encompass
The Intact Genomics IG RNA Cleanup Kit purifies up to 50 µg of RNA. Has a rapid and simplified workflow with a single wash buffer. Efficiently purify RNA ≥ 25 nt (a simple modification enables purification of RNA ≥ 15 nt;)Option to Elute in 20-50 µL for more concentrated RNA than the standard 100 µL; Standard 70-100% RNA recovery; Customizable buffer options for difficult reactions; Purified RNA is ready for use in a wide variety of downstream applications including: RT-PCR, RNA library prep for NGS, formation of RNP complexes for genome editing, microinjection, RNA labeling, and transfection.This kit is recommended for use following the IG sgRNA Synthesis Kits to remove proteins, salts and most unincorporated nucleotides. The Intact Genomics RNA Cleanup Kit has a simplified workflow and is ideal for cleanup and concentration of RNA after enzymatic treatments such as DNase I, Proteinase K, labeling, capping or in vitro transcription (IVT).
Catalog No. 50-000-83005 Supplier INTACT GENOMICS INC Supplier No. 4005
The Intact Genomics IG RNA Cleanup Kit purifies up to 50 µg of RNA. Has a rapid and simplified workflow with a single wash buffer. Efficiently purify RNA ≥ 25 nt (a simple modification enables purification of RNA ≥ 15 nt;)Option to Elute in 20-50 µL for more concentrated RNA than the standard 100 µL; Standard 70-100% RNA recovery; Customizable buffer options for difficult reactions; Purified RNA is ready for use in a wide variety of downstream applications including: RT-PCR, RNA library prep for NGS, formation of RNP complexes for genome editing, microinjection, RNA labeling, and transfection.This kit is recommended for use following the IG sgRNA Synthesis Kits to remove proteins, salts and most unincorporated nucleotides. The Intact Genomics RNA Cleanup Kit has a simplified workflow and is ideal for cleanup and concentration of RNA after enzymatic treatments such as DNase I, Proteinase K, labeling, capping or in vitro transcription (IVT).