Learn More
Boca Scientific Inc GeneFix RNA Saliva Collectors, Pre-filled with Stabilization Buffer, Pack of 50
Catalog No. 501685615
Encompass
GeneFix RNA saliva collectors are easy to use saliva collection devices that stabilize RNA in tubes pre-filled with a non-toxic stabilization buffer. The collector has a simple screw-on funnel for saliva delivery directly into the buffer. The saliva collection tube clearly indicates the volume of saliva required. The RNA is stabilized in the buffer for 2 weeks at room temperature, with high yields maintained. GeneFix saliva collectors are fully specified for sample transport and storage.
Catalog No. 50-168-5615 Supplier Boca Scientific Inc Supplier No. RFX01/50
GeneFix RNA saliva collectors are easy to use saliva collection devices that stabilize RNA in tubes pre-filled with a non-toxic stabilization buffer. The collector has a simple screw-on funnel for saliva delivery directly into the buffer. The saliva collection tube clearly indicates the volume of saliva required. The RNA is stabilized in the buffer for 2 weeks at room temperature, with high yields maintained. GeneFix saliva collectors are fully specified for sample transport and storage.