Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ RNA (-) Control CISH Probe

Catalog No. 89335571 Shop All Abnova Corporation Products
Change view
Click to view available options
For Use With (Application):
CISH-P
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
89-335-571 CISH-P
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-571 Supplier Abnova™ Supplier No. CO0002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RNA (-) Control CISH Probe is designed to be used for assessing the unspecific background staining in formalin-fixed, paraffin-embedded tissue or cells by chromogenic in situ hybridization (CISH).

The probe is intended to be used in combination with the CISH Implementation HRP-DAB Kit, which provides necessary reagents for specimen pretreatment and post-hybridization processing.

This probe consists of a set of random sequence oligonucleotides with GC contents of 40-70% without known consensus to any naturally occurring sequences. This probe should not result in positive staining signals and is to be used to assess the unspecific background staining within specimens. Visualization of signals should be performed by light microscopy using a 10x or 20x objective. For signal evaluation, necrotic, degenerated or over-digested cells should be avoided as these cells often stain nonspecifically. In order to judge the specificity of the hybridization signals and to confirm the correct performance of the method, any hybridization should be accompanied by controls. We recommend using at least one control sample containing both true positive and negative staining cells.

Specifications

Content And Storage Bring probe to hybridization temperature before use.
For Use With (Application) CISH-P
Includes Reagent Provided:

Digoxigenin-labeled oligonucleotides with GC contents of 40-70% without known consensus to any naturally occurring sequences.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.