Learn More
Bioworld RNA-DNA Spin Columns w/ Glass Fiber Disk, 100 Column
Supplier:  Bioworld 211400132
Encompass_Preferred
Mini Spin columns for RNA/DNA extraction and purification.Specifications3 layered glass fiber disk Pore size: 1μmSample size: 750μlBinding capacity: 20μg DNA/100μg RNAApplications:Agarose gel extractionPlasmid isolation PCR clean upPlamid isolation
Catalog No. 50-198-568