Learn More
Bioworld RNA Electrophoresis Kit, 1 Kit
Catalog No. 50196419
Encompass_Preferred
RNA electrophoresis kit contains all the necessary reagents packaged conveniently as a kit for gel electrophoresis of RNA.Kit components:Agarose I: 10g6X Agarose gel loading dye: 500μlMOPS 10x liquid concentrate: 100mlFormamide deionized: 10mlEthidium Bromide (625μg/ml) dropper bottle: 5mlNuclease Eliminater wipes 3pksDestainer bags: 3
Catalog No. 50-196-419 Supplier Bioworld Supplier No. 104200001
RNA electrophoresis kit contains all the necessary reagents packaged conveniently as a kit for gel electrophoresis of RNA.Kit components:Agarose I: 10g6X Agarose gel loading dye: 500μlMOPS 10x liquid concentrate: 100mlFormamide deionized: 10mlEthidium Bromide (625μg/ml) dropper bottle: 5mlNuclease Eliminater wipes 3pksDestainer bags: 3