Learn More
Biochain Institute Inc Whole Blood RNA Extraction Kit, 1 Kit/PK
SDP
Catalog No. 50183151
Encompass_Preferred
BioChain's Whole Blood RNA Isolation Kit provides a rapid method for the isolation and purification of RNA molecules from 200 ul whole blood samples. The RNA molecules isolated using BioChain's Whole Blood Isolation Kit can be used in various downstream applications relating to gene regulation and functional analysis, including qRT-PCR, northern blotting and microarray profiling analysis.
Catalog No. 50-183-151 Supplier Biochain Institute Inc Supplier No. K1012050
BioChain's Whole Blood RNA Isolation Kit provides a rapid method for the isolation and purification of RNA molecules from 200 ul whole blood samples. The RNA molecules isolated using BioChain's Whole Blood Isolation Kit can be used in various downstream applications relating to gene regulation and functional analysis, including qRT-PCR, northern blotting and microarray profiling analysis.