Learn More
Biochain Institute Inc RNAmate (Polysaccharide and ProteoglySAN-Rh-Cy Decontamination Solution), 100 ml/PK
SDP
Catalog No. 50183149
Encompass_Preferred
A common problem in current RNA isolation methods is the contamination of RNA with polysaccharides and proteoglycans, which will adversely affect migration of RNA in gel electrophoresis and inhibit the reverse transcription -polymerase chain reaction (RT-PCR). RNAmate is a ready to use reagent for decontamination of polysaccharides and proteoglycans from RNA preparations, and our testing shows that RNAmate increases the purity of total RNA. RNAmate is the only polysaccharide and proteoglycan decontamination solution for RNA purification on the market.
Catalog No. 50-183-149 Supplier Biochain Institute Inc Supplier No. L1011100
A common problem in current RNA isolation methods is the contamination of RNA with polysaccharides and proteoglycans, which will adversely affect migration of RNA in gel electrophoresis and inhibit the reverse transcription -polymerase chain reaction (RT-PCR). RNAmate is a ready to use reagent for decontamination of polysaccharides and proteoglycans from RNA preparations, and our testing shows that RNAmate increases the purity of total RNA. RNAmate is the only polysaccharide and proteoglycan decontamination solution for RNA purification on the market.