Learn More
Biochain Institute Inc Total RNA Extraction Kit, 1 kit/PK
SDP
Catalog No. 50183150
Encompass_Preferred
BioChain's Total RNA Extraction kit is a convenient tool for isolating high quality Total RNAs. The isolated Total RNAs can be used for mRNA isolation, probe generation, RT-PCR, Northern blot analysis, primer extension, RNA protection assay, and In vitro translation etc.. The kit contains enough reagents for isolating Total RNAs from 5 gram tissue and cells.
Catalog No. 50-183-150 Supplier Biochain Institute Inc Supplier No. K2014005
BioChain's Total RNA Extraction kit is a convenient tool for isolating high quality Total RNAs. The isolated Total RNAs can be used for mRNA isolation, probe generation, RT-PCR, Northern blot analysis, primer extension, RNA protection assay, and In vitro translation etc.. The kit contains enough reagents for isolating Total RNAs from 5 gram tissue and cells.