Learn More
Biochain Institute Inc FFPE Tissue RNA Extraction Kit, 1 kit/PK
SDP
Catalog No. 50183146
Encompass_Preferred
BioChain's FFPE Tissue RNA Extraction Kit allows for facile and efficient ribonucleic acid extraction from FFPE tissues. All necessary reagents for RNA extractions in FFPE tissue specimens are provided, including FFPE tissue lysis buffer, proteinase K, and an actin control primer for control RT-qPCR reactions. The kit contains sufficient reagents for 100 FFPE tissue RNA extraction reactions.
Catalog No. 50-183-146 Supplier Biochain Institute Inc Supplier No. K1011100
BioChain's FFPE Tissue RNA Extraction Kit allows for facile and efficient ribonucleic acid extraction from FFPE tissues. All necessary reagents for RNA extractions in FFPE tissue specimens are provided, including FFPE tissue lysis buffer, proteinase K, and an actin control primer for control RT-qPCR reactions. The kit contains sufficient reagents for 100 FFPE tissue RNA extraction reactions.