Learn More
Biochain Institute Inc MicroRNA Isolation Kit, 1 kit/PK
SDP
Catalog No. 50183148
Encompass_Preferred
BioChain's MicroRNA Isolation Kit provides a rapid method for the isolation and purification of small RNA molecules (<200 nt) not only from tissue culture cells and tissues, but also from blood and serum. This kit can also be used for isolating large RNA (>200 nt). This kit contains enough reagents for 25 isolations of small RNAs (<200 nt) or 50 isolations of large RNAs (>200 nt).
Catalog No. 50-183-148 Supplier Biochain Institute Inc Supplier No. KS341025
BioChain's MicroRNA Isolation Kit provides a rapid method for the isolation and purification of small RNA molecules (<200 nt) not only from tissue culture cells and tissues, but also from blood and serum. This kit can also be used for isolating large RNA (>200 nt). This kit contains enough reagents for 25 isolations of small RNAs (<200 nt) or 50 isolations of large RNAs (>200 nt).