Learn More
Omega Biotek Inc E.Z.N.A. HP Total RNA Kit, 200 preps, RNA binding capacity 100 μg (RNA >200 bases), silica-based spin column
Catalog No. NC1711531
Encompass
The OMEGA BIOTEK E.Z.N.A. HP Total RNA Kit provides an efficient solution for the isolation of high-quality total RNA from eukaryotic cells and tissues. It employs a streamlined purification process, integrating sample homogenization and genomic DNA removal into a single step, utilizing advanced column technology. This kit enhances user safety by eliminating toxic organic solvents and simplifies the protocol for RNA purification, making it suitable for various sensitive applications.
  • Rapid isolation in approximately 25 minutes for quick experimental turnaround
  • High-performance RNA quality and yield
  • Versatile compatibility with various cell and tissue types
  • Safe and convenient with no toxic organic solvents
  • Optimized for sensitive downstream applications like RT-PCR and next-generation sequencing
  • High binding capacity of 100 μg for RNA >200 bases
Catalog No. NC1711531 Supplier Omega Biotek Inc Supplier No. R681202
The OMEGA BIOTEK E.Z.N.A. HP Total RNA Kit provides an efficient solution for the isolation of high-quality total RNA from eukaryotic cells and tissues. It employs a streamlined purification process, integrating sample homogenization and genomic DNA removal into a single step, utilizing advanced column technology. This kit enhances user safety by eliminating toxic organic solvents and simplifies the protocol for RNA purification, making it suitable for various sensitive applications.
  • Rapid isolation in approximately 25 minutes for quick experimental turnaround
  • High-performance RNA quality and yield
  • Versatile compatibility with various cell and tissue types
  • Safe and convenient with no toxic organic solvents
  • Optimized for sensitive downstream applications like RT-PCR and next-generation sequencing
  • High binding capacity of 100 μg for RNA >200 bases