Learn More
Alkali Scientific RNA Extraction Spin Columns 2m
SDP
Catalog No. 503027650
Encompass_Preferred
Alkali ScientificRNA Spin Columns provide a fast efficient and reliable method for extracting high-quality RNA from a variety of samples including tissues cells plants and blood Engineered with a robust polypropylene construction these columns ensure chemical resistance and sample integrity while their advanced silica membrane technology enables efficient RNA binding washing and elution Each unit is supplied with collection tubes for seamless processing in standard centrifuges These columns deliver highly pure RNA with reproducible yields suitable for downstream applications such as RT-PCR qPCR RNA sequencing and gene expression studies The DNase- and RNase-free construction ensures sample integrity while human DNA-free certification supports applications requiring high sensitivity and specificity Polypropylene Construction Durable and chemically compatible for reliable performance High Purity RNA Extracts intact RNA with minimal degradation ideal for advanced molecular applicati
Catalog No. 50-302-7650 Supplier Alkali Scientific Supplier No. GE8002
Alkali ScientificRNA Spin Columns provide a fast efficient and reliable method for extracting high-quality RNA from a variety of samples including tissues cells plants and blood Engineered with a robust polypropylene construction these columns ensure chemical resistance and sample integrity while their advanced silica membrane technology enables efficient RNA binding washing and elution Each unit is supplied with collection tubes for seamless processing in standard centrifuges These columns deliver highly pure RNA with reproducible yields suitable for downstream applications such as RT-PCR qPCR RNA sequencing and gene expression studies The DNase- and RNase-free construction ensures sample integrity while human DNA-free certification supports applications requiring high sensitivity and specificity Polypropylene Construction Durable and chemically compatible for reliable performance High Purity RNA Extracts intact RNA with minimal degradation ideal for advanced molecular applicati