Learn More
MyBioSource RNA (For fixed/embedded tissues)(without Phenol & Chloroform, need not DNase I digestion, directly use in RT-PCR or qPCR) Isolation Kit, 50 Tests
SDP
Catalog No. NC3876613
Encompass_Preferred
This kit is designed to quickly extract total RNA from formalin-fixed and paraffin-embedded tissue samples. The unique lysis solution/proteinase K quickly lyses cells to release RNA, and then the lysis mixture passes through a genomic DNA removal column, where genomic DNA is removed and RNA is filtered through. After the filtered RNA is adjusted with ethanol to adjust the binding conditions, the RNA is selectively adsorbed on the silicon matrix membrane in the spin column in a high-isolated salt state, and then through a series of rapid rinsing-centrifugation steps, the protein-removing solution and the rinsing solution remove the cells. Metabolites, proteins and other impurities are removed, and finally the low-salt RNase free H20 will elute the pure RNA from the silicon matrix membrane. The obtained RNA can be used in experiments such as reverse transcription PCR and fluorescent quantitative PCR.
Catalog No. NC3876613 Supplier MyBioSource Supplier No. MBS35595650 TESTS
This kit is designed to quickly extract total RNA from formalin-fixed and paraffin-embedded tissue samples. The unique lysis solution/proteinase K quickly lyses cells to release RNA, and then the lysis mixture passes through a genomic DNA removal column, where genomic DNA is removed and RNA is filtered through. After the filtered RNA is adjusted with ethanol to adjust the binding conditions, the RNA is selectively adsorbed on the silicon matrix membrane in the spin column in a high-isolated salt state, and then through a series of rapid rinsing-centrifugation steps, the protein-removing solution and the rinsing solution remove the cells. Metabolites, proteins and other impurities are removed, and finally the low-salt RNase free H20 will elute the pure RNA from the silicon matrix membrane. The obtained RNA can be used in experiments such as reverse transcription PCR and fluorescent quantitative PCR.