Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc RNA Gel Buffer (10X) (MOPS Buffer), 500 mL
SDP

Catalog No. 50983261
Encompass_Preferred

MOPS Buffer is used at 1X for the electrophoresis of RNA as the running buffer to separate RNA samples on agarose and formaldehyde-agarose (denaturing) gels. MOPS buffer is also used for Northern Blots -the hybridization of total RNA or mRNA to a membrane. Liquid, 10X concentration

Filtered through a 0.22 µm membrane
DNase, RNase Tested

Applications: MOPS Buffer is suitable for use in RNA Gel Electrophoresis, Northern Blot Analysis, and when making formaldehyde-agarose gels and associated running buffer for RNA electrophoresis.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
DNase, RNase Tested
pH: 7.0 ± 0.1
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-983-261 Supplier Quality Biological Inc Supplier No. 351059101
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MOPS Buffer is used at 1X for the electrophoresis of RNA as the running buffer to separate RNA samples on agarose and formaldehyde-agarose (denaturing) gels. MOPS buffer is also used for Northern Blots -the hybridization of total RNA or mRNA to a membrane. Liquid, 10X concentration

Filtered through a 0.22 µm membrane
DNase, RNase Tested

Applications: MOPS Buffer is suitable for use in RNA Gel Electrophoresis, Northern Blot Analysis, and when making formaldehyde-agarose gels and associated running buffer for RNA electrophoresis.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
DNase, RNase Tested
pH: 7.0 ± 0.1
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.