Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc RNA Gel Loading Solution (10X), 10 mL
SDP

Catalog No. 507517246
Encompass_Preferred

RNA Gel Loading Solution, 10X is prepared according to Sambrook, Fritsch & Maniatis (1989). The purpose of this reagent is to insure that the RNA samples remain submerged in the wells of the gel, by making them denser than the electrophoresis buffer.

Contains two dyes, bromphenol blue & xylene cyanole FF
Filtered through a 0.22 µm membrane
RNase tested

Applications: RNA Gel Loading Solution is suitable for RNA Gel Electrophoresis and Northern Blot Analysis.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
RNase Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-751-7246 Supplier Quality Biological Inc Supplier No. 351081661EA
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RNA Gel Loading Solution, 10X is prepared according to Sambrook, Fritsch & Maniatis (1989). The purpose of this reagent is to insure that the RNA samples remain submerged in the wells of the gel, by making them denser than the electrophoresis buffer.

Contains two dyes, bromphenol blue & xylene cyanole FF
Filtered through a 0.22 µm membrane
RNase tested

Applications: RNA Gel Loading Solution is suitable for RNA Gel Electrophoresis and Northern Blot Analysis.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
RNase Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.