Learn More
Apexbio Technology LLC RNA LIB PREP KIT PREMIX-24mg
SDP
Catalog No. 5000897364
Encompass_Preferred
RNA Library Prep Kit (Premixed Version) is essential in genomic research facilitating RNA sequencing by preparing cDNA libraries from RNA samples Originating from advancements in molecular biology it targets RNA molecules to enable comprehensive transcriptional profiling This kit operates by converting RNA to cDNA employing reverse transcription and amplification pathways which are pivotal for accurate gene expression analysis It exhibits high efficacy across various concentrations often in nanomolar to low micromolar ranges Widely used in cell models animal studies and drug screening this kit is integral for exploring genetic variations and expressions Experimental concentrations typically depend on the specific research design
Catalog No. 50-008-97364 Supplier Apexbio Technology LLC Supplier No. K181024
RNA Library Prep Kit (Premixed Version) is essential in genomic research facilitating RNA sequencing by preparing cDNA libraries from RNA samples Originating from advancements in molecular biology it targets RNA molecules to enable comprehensive transcriptional profiling This kit operates by converting RNA to cDNA employing reverse transcription and amplification pathways which are pivotal for accurate gene expression analysis It exhibits high efficacy across various concentrations often in nanomolar to low micromolar ranges Widely used in cell models animal studies and drug screening this kit is integral for exploring genetic variations and expressions Experimental concentrations typically depend on the specific research design