Learn More
Bioworld RNA Molecular Weight Marker, 16S + 23S Ribosomal RNA (1776-3566 bp), 500 μg
Supplier:  Bioworld 104300131
Encompass_Preferred
- Ultra Pure Grade - Ribosomal RNA, Purified from Ecoli, consisting of 2 fragments approximately 1,776bp and 3,566bp in length.Function Test(P/F)- Electrophoresis Analysis, Pass RNase (P/F)- None Detected
Catalog No. 50-196-431