Learn More
Takara Bio RNA-OFF 500ML
SDP
Catalog No. 50443879
Encompass
Solution, Decontamination; RNase-Off; Stable and heat resistant; Ready to use; Works in less than 1 min.; Safe on lab surfaces, plastic, glass, and pipetters; 16 oz. (500mL)
Catalog No. 50-443-879 Supplier Takara Bio Supplier No. 9037
Solution, Decontamination; RNase-Off; Stable and heat resistant; Ready to use; Works in less than 1 min.; Safe on lab surfaces, plastic, glass, and pipetters; 16 oz. (500mL)