Learn More
Enzo Life Sciences RNA polymerase II monoclonal antibody (H14) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS134A0100
Encompass_Preferred
Clone: H14. Isotype: IgM. Immunogen: Purified phosphorylated form of RNA polymerase II extracted from a transformed cell line. Species reactivity: Species independent. Specificity: Recognizes RNA polymerase II phosphorylated at Ser5 in a broad range of species (from yeast to human). Applications: IF, IP, WB. Purity: Purified by affinity chromatography. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide.
Catalog No. 50-201-2697