Learn More
Abcam Mouse monoclonal [4H8] to RNA polymerase II CTD repeat YSPTSPS (phospho S5) - ChIP Grade
Supplier: Abcam AB5408100UG
Mouse monoclonal [4H8] to RNA polymerase II CTD repeat YSPTSPS (phospho S5) - ChIP Grade
The product is subject to the following: Abcam Restricted Use Statement