Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RNA Polymerase beta Antibody (YA2403) | 10 UL
SDP

Catalog No. 5000445057
Encompass_Preferred

RNA Polymerase beta Antibody (YA2403) is a rabbit-derived, non-conjugated IgG monoclonal antibody, specifically targeting RNA Polymerase beta. This antibody is validated for Western Blot (WB) and Immunoprecipitation (IP) applications. RNA Polymerase beta itself is a DNA-dependent RNA polymerase (RNAP) essential for transcribing DNA into RNA, and mutations in this protein are known to confer resistance to various antibiotics.

  • Host: Rabbit
  • Reactivity: E.coli
  • Application: WB, IP
  • Clonality: Monoclonal
  • Immunogen: RNA Polymerase beta
  • Conjugation: Non-conjugated
  • Isotype: IgG
  • Appearance: Liquid
  • Shipping: With blue ice
  • Storage: Store at -20°C for 1 year, avoid repeated freeze/thaw cycles.

Catalog No. 50-004-45057 Supplier Medchemexpress LLC Supplier No. HYP8265810UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RNA Polymerase beta Antibody (YA2403) is a rabbit-derived, non-conjugated IgG monoclonal antibody, specifically targeting RNA Polymerase beta. This antibody is validated for Western Blot (WB) and Immunoprecipitation (IP) applications. RNA Polymerase beta itself is a DNA-dependent RNA polymerase (RNAP) essential for transcribing DNA into RNA, and mutations in this protein are known to confer resistance to various antibiotics.

  • Host: Rabbit
  • Reactivity: E.coli
  • Application: WB, IP
  • Clonality: Monoclonal
  • Immunogen: RNA Polymerase beta
  • Conjugation: Non-conjugated
  • Isotype: IgG
  • Appearance: Liquid
  • Shipping: With blue ice
  • Storage: Store at -20°C for 1 year, avoid repeated freeze/thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.