Learn More
Bioss RNA polymerase beta Recombinan
SDP
Catalog No. 5000065824
Encompass_Preferred
RNA polymerase beta Primary Antibody Unconjugated Host Species Rabbit Target Species E coli Clonality Monoclonal Applications WB IP RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-65824 Supplier Bioss Supplier No. BSM61640R
RNA polymerase beta Primary Antibody Unconjugated Host Species Rabbit Target Species E coli Clonality Monoclonal Applications WB IP RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol