Learn More
Medchemexpress LLC RNA Polymerase II Subunit B1 Antibody (YA090) | 10 UL
SDP
Catalog No. 5000441982
Encompass_Preferred
RNA Polymerase II Subunit B1 Antibody (YA090) is a rabbit-derived and non-conjugated IgG monoclonal antibody, targeting RNA Polymerase II Subunit B1. This gene encodes the largest subunit of RNA polymerase II, responsible for synthesizing messenger RNA in eukaryotes. The product contains a carboxy terminal domain of heptapeptide repeats, essential for polymerase activity. These repeats have serine and threonine residues that are phosphorylated in actively transcribing RNA polymerase. This subunit, with other polymerase subunits, forms the DNA binding domain, a groove where the DNA template is transcribed into RNA.
  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Targets RNA Polymerase II Subunit B1
  • Reactive with human samples
  • Applications include WB (1:500-1:1000), IHC-P (1:50-1:100), IHC-F (1:50-1:100), ICC/IF (1:50-1:200), and IP (1:20)
Catalog No. 50-004-41982 Supplier Medchemexpress LLC Supplier No. HYP8088310UL
RNA Polymerase II Subunit B1 Antibody (YA090) is a rabbit-derived and non-conjugated IgG monoclonal antibody, targeting RNA Polymerase II Subunit B1. This gene encodes the largest subunit of RNA polymerase II, responsible for synthesizing messenger RNA in eukaryotes. The product contains a carboxy terminal domain of heptapeptide repeats, essential for polymerase activity. These repeats have serine and threonine residues that are phosphorylated in actively transcribing RNA polymerase. This subunit, with other polymerase subunits, forms the DNA binding domain, a groove where the DNA template is transcribed into RNA.
  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Targets RNA Polymerase II Subunit B1
  • Reactive with human samples
  • Applications include WB (1:500-1:1000), IHC-P (1:50-1:100), IHC-F (1:50-1:100), ICC/IF (1:50-1:200), and IP (1:20)