Learn More
VAZYME INTERNATIONAL LLC Vazyme VAHTS Universal V10 RNA-seq Library Prep Kit, Premixed Version
Catalog No. NC3979702
Encompass
The VAHTS Universal V10 RNA-seq Library Prep Kit (Premixed Version) is a transcriptome library preparation kit specifically developed for high-throughput sequencing platforms. It contains premixed reagents to simplify operations and reduce hands-on time, allowing for the preparation of non-stranded or stranded transcriptome libraries. This kit is compatible with multiple species and delivers a high library yield.
  • Simplified operation with premixed reagents enables completion in just 4 hours.
  • Efficiently works with a diverse range of species.
  • Supports leading high-throughput sequencing platforms.
  • Achieves high mapping and mRNA ratios compared to other suppliers.
  • Can prepare non-stranded or stranded transcriptome libraries as needed.
Catalog No. NC3979702 Supplier VAZYME INTERNATIONAL LLC Supplier No. NR61601
The VAHTS Universal V10 RNA-seq Library Prep Kit (Premixed Version) is a transcriptome library preparation kit specifically developed for high-throughput sequencing platforms. It contains premixed reagents to simplify operations and reduce hands-on time, allowing for the preparation of non-stranded or stranded transcriptome libraries. This kit is compatible with multiple species and delivers a high library yield.
  • Simplified operation with premixed reagents enables completion in just 4 hours.
  • Efficiently works with a diverse range of species.
  • Supports leading high-throughput sequencing platforms.
  • Achieves high mapping and mRNA ratios compared to other suppliers.
  • Can prepare non-stranded or stranded transcriptome libraries as needed.