Learn More
Apexbio Technology LLC RNA Stabilization Solution for Animal Tissue 100mL
SDP
Catalog No. 5000097226
Encompass_Preferred
RNA Stabilization Solution for Animal Tissue is a reagent designed to stabilize RNA within animal tissues and cells It works by rapidly penetrating tissue and effectively inhibiting RNase enzymes thereby preventing RNA degradation This enables preservation of RNA integrity at ambient temperatures post-collection eliminating the need for immediate snap-freezing in liquid nitrogen Based on these properties RNA Stabilization Solution for Animal Tissue is suitable for preserving RNA expression profiles in animal tissue samples during collection transport and storage supporting downstream applications such as transcriptome analysis and quantitative gene expression studies
Catalog No. 50-000-97226 Supplier Apexbio Technology LLC Supplier No. K1624100
RNA Stabilization Solution for Animal Tissue is a reagent designed to stabilize RNA within animal tissues and cells It works by rapidly penetrating tissue and effectively inhibiting RNase enzymes thereby preventing RNA degradation This enables preservation of RNA integrity at ambient temperatures post-collection eliminating the need for immediate snap-freezing in liquid nitrogen Based on these properties RNA Stabilization Solution for Animal Tissue is suitable for preserving RNA expression profiles in animal tissue samples during collection transport and storage supporting downstream applications such as transcriptome analysis and quantitative gene expression studies