Learn More
Active Motif RNA Subcellular Isolation Kit
SDP
Catalog No. NC1841577
Encompass
Active Motif's RNA Subcellular Isolation Kit provides an efficient method to isolate nuclear and cytoplasmic RNA without cross-contamination. It can be used to isolate both short and long RNA sequences (e.g. miRNA, piRNA, snoRNA, mRNA, and lncRNA). RNA molecules exist at different levels of maturation and processing within a cell at any given time. Such heterogeneity can bias downstream analysis and interpretation of data. By isolating RNA from specific subcellular fractions, the accuracy of gene expression analysis can be greatly improved. Subcellular isolation enhances the ability to detect low abundance transcripts and aids in the ability to study RNA processing dynamics by reducing the background from other intronic or mature RNAs. Additionally, subcellular isolation helps […] For more information, please visit: https://www.activemotif.com/catalog/1160.
Catalog No. NC1841577 Supplier Active Motif Supplier No. 25501
Active Motif's RNA Subcellular Isolation Kit provides an efficient method to isolate nuclear and cytoplasmic RNA without cross-contamination. It can be used to isolate both short and long RNA sequences (e.g. miRNA, piRNA, snoRNA, mRNA, and lncRNA). RNA molecules exist at different levels of maturation and processing within a cell at any given time. Such heterogeneity can bias downstream analysis and interpretation of data. By isolating RNA from specific subcellular fractions, the accuracy of gene expression analysis can be greatly improved. Subcellular isolation enhances the ability to detect low abundance transcripts and aids in the ability to study RNA processing dynamics by reducing the background from other intronic or mature RNAs. Additionally, subcellular isolation helps […] For more information, please visit: https://www.activemotif.com/catalog/1160.