Learn More
Sigma Aldrich Fine Chemicals Biosciences RNA ZAP CASE OF 6X 250 ML
Catalog No. NC0771205
Encompass
RNASE ZAP 6x250ml. A cleaning agent for removing RNase from glassware, plastic surfaces, countertops, and pipettors. It is also effective at eliminating RNase contamination from microcentrifuge tubes without inhibiting subsequent enzymatic reactions.
Catalog No. NC0771205 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R20206X250ML
RNASE ZAP 6x250ml. A cleaning agent for removing RNase from glassware, plastic surfaces, countertops, and pipettors. It is also effective at eliminating RNase contamination from microcentrifuge tubes without inhibiting subsequent enzymatic reactions.