Learn More
Cayman Chemical RnaconItIn 10mg
SDP
Catalog No. 502910455
Encompass_Preferred
An alkaloid with analgesic and anesthetic activities (ED50s = 6.99 and 0.167 µmol/kg, respectively, in mice)
Catalog No. 50-291-0455 Supplier Cayman Chemical Supplier No. 3477410 MG
An alkaloid with analgesic and anesthetic activities (ED50s = 6.99 and 0.167 µmol/kg, respectively, in mice)