Learn More
Sigma Aldrich Fine Chemicals Biosciences RNAlaterR Stabilize and pr
Catalog No. 502901572
Encompass
RNAlater is compatible with most RNA isolation methods including TRI Reagent and GenElute Total RNA isolation and mammalian mRNA isolation kits.RNAlater is an aqueous non-toxic tissue storage reagent that rapidly permeates tissue to stabilize and protect cellular RNA in situ in unfrozen specimens. Tissue pieces are harvested and immediately submerged in RNAlater for storage without jeopardizing the quality or quantity of RNA. RNAlater eliminates the need to immediately process tissue specimens or to freeze samples in liquid nitrogen for later processing. RNAlater preserves RNA in tissues for up to 1 day at 37 C 1 week at 25 C and 1 month at 4 C. Tissues can also be stored at -20 C long-term.RNAlater has been extensively tested on several tissues from vertebrate species including brain heart kidney spleen liver testis skeletal muscle fat lung and thymus. RNAlater is also effective for E. coli Drosophila tissue culture cells white blood cells and some plants.
Catalog No. 50-290-1572 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R0901100ML
RNAlater is compatible with most RNA isolation methods including TRI Reagent and GenElute Total RNA isolation and mammalian mRNA isolation kits.RNAlater is an aqueous non-toxic tissue storage reagent that rapidly permeates tissue to stabilize and protect cellular RNA in situ in unfrozen specimens. Tissue pieces are harvested and immediately submerged in RNAlater for storage without jeopardizing the quality or quantity of RNA. RNAlater eliminates the need to immediately process tissue specimens or to freeze samples in liquid nitrogen for later processing. RNAlater preserves RNA in tissues for up to 1 day at 37 C 1 week at 25 C and 1 month at 4 C. Tissues can also be stored at -20 C long-term.RNAlater has been extensively tested on several tissues from vertebrate species including brain heart kidney spleen liver testis skeletal muscle fat lung and thymus. RNAlater is also effective for E. coli Drosophila tissue culture cells white blood cells and some plants.