Learn More
Enzo Life Sciences RNA polymerase II monoclonal antibody (8WG16) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS132A0100
Encompass_Preferred
Clone: 8WG16. Isotype: IgG2a. Immunogen: RNA polymerase (RNA pol II) purified from wheat germ extract. Species reactivity: Species independent. Specificity: Recognizes RNA polymerase II in a broad range of species (from yeast to human). Detects the C-terminal heptapeptide repeat present on the largest subunit of pol II. Applications: IF, IP, WB. Purity: Affinity purified. Formulation: Liquid. In PBS, pH 7.2.
Catalog No. 50-201-2696